Anti-CCND1 Mouse Monoclonal Antibody
Supplier: Enzo Life Sciences
905-776-100
95038-974EA 395.67 USD
95038-974
Anti-CCND1 Mouse Monoclonal Antibody
Antibodies
Cyclins are a family of proteins which function as the regulatory subunits of cyclin/cyclin-dependent kinase (Cdk) holoenzymes, mediating entry into and progression through the cell cycle. Each cyclin is expressed in a temporal fashion during the cell cycle, partnering with a specific Cdk to activate its protein kinase activity. The most well defined mitotic cyclins are: cyclin D, which partners with cdks 4 and 6 during early G1 phase; cyclin E, which activates Cdk2 in the G1 to S phase transition; cyclin A, which utilizes Cdks 1 and 2 during progression through S phase and into G2; andMore Product Information