Fluorescein-12-ddUTP, Enzo Life Sciences
Supplier: Enzo Life Sciences
ENZ-42833
89165-844EA 462.16 USD
89165-844
Fluorescein-12-ddUTP, Enzo Life Sciences
Nucleic Acid Reagents Nucleotides
Fluorescein-12-ddUTP can be used for 3’-end labeling of oligodeoxynucleotides using terminal transferase. It also serves as a substrate for E. coli DNA Polymerase I (holoenzyme and Klenow fragment), T4 and T7 DNA polymerases, Taq DNA polymerase and reverse transcriptase. Oligonucleotide probes labeled with fluorescein-12-ddUTP can be detected by direct fluorescence detection (green fluorescence) or by methods using horseradish peroxidase- or alkaline phosphatase-linked anti-fluorescein antibodies.