Anti-TRPV1 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
BML-SA556-0100
89163-310EA 773.9 USD
89163-310
Anti-TRPV1 Rabbit Polyclonal Antibody
Antibodies
Host: Rabbit

  • For Immunocytochemistry, Immunohistochemistry

Recognizes human TRPV1.

Type: Primary
Antigen: TRPV1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR