Anti-GABRA1 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
BML-SA512-0100
89163-224EA 577.7 USD
89163-224
Anti-GABRA1 Rabbit Polyclonal Antibody
Antibodies
Host: Rabbit

  • For Western Blot

Recognizes human, mouse and rat GABA A receptor (alpha1-subunit).

Type: Primary
Antigen: GABA A RECEPTOR ALPHA 1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR