Anti-ADAM15 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
BML-SA428-0100
89163-060EA 382.59 USD
89163-060
Anti-ADAM15 Rabbit Polyclonal Antibody
Antibodies
Host: Rabbit

  • For Western Blot

Recognizes human, rat, mouse and other mammalian ADAM15.

Type: Primary
Antigen: ADAM15
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR