Anti-Proteasome 19S Rpt5/S6A Subunit Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
PW8245-0025 PW8245-0100
89162-092EA 122.08 USD
89162-092 89162-094
Anti-Proteasome 19S Rpt5/S6A Subunit Rabbit Polyclonal Antibody
Antibodies
Host: Rabbit

  • For Western Blot

Recognizes yeast proteasome 26S (19S regulator complex) (Rpt5/S6a subunit).

Type: Primary
Antigen: PROTEASOME 19S RPT5/S6A SUBUNIT
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Yeast
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR