Anandamide (18:2, n-6) ≥98%

Supplier: Enzo Life Sciences

Synonyms: Linoleyl ethanolamide, (9Z,12Z)-N-(2-Hydroxyethyl)octadeca-9,12-dienamide, Linoleylethanolamide, N-(2-Hydroxyethyl)linoleamide, n-linoleoylethanolamine, Linoleamide MEA, Linoleic ethanolamide

BML-FA026-0005
89158-390EA 86.11 USD
89158-390
Anandamide (18:2, n-6) ≥98%
Anandamide (18:2, n-6)

Endocannabinoid.


  • Formulation: Waxy solid.


Anandamide analog.


Soluble in ethanol or DMSO.

Formula: C₂₀H₃₇NO₂
MW: 323.5 g/mol
Storage Temperature: Ultra-Low Temperature Freezer (–80°C)
CAS Number: 68171-52-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fa94fa02a9518722109026fa3be27fd8f93f394f&vid=98cb7717c82013a0ae9dd8c825d8e08ce14d9c2e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥98% (TLC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR