G418 disulfate (from Micromonospora rhodorangea)

Supplier: Enzo Life Sciences

Synonyms: G418 disulfate, G-418 sulfate, G-418 sulphate, Geneticin®, G-418 disulphate, G418 sulphate, G-418 disulfate

380-013-G001 380-013-G005 380-013-M500
89149-806EA 69.76 USD
89149-806 89149-808 89149-812
G418 disulfate (from Micromonospora rhodorangea)
G418 disulphate

White to off-white powder


Gentamycin-class aminoglycoside antibiotic. Cytotoxic to prokaryotic and eukaryotic cells. Induces apoptosis. Widely employed in the selection of eukaryotic expression vectors, in combination with either aminoglycoside phosphotransferase 3' or APH II.


Soluble in water


Packaging: Shipping temperature: Ambient

Formula: C₂₀H₄₄N₄O₁₈S₂
MW: 692.72 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00058314
CAS Number: 108321-42-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b829ba286bff8e88aee0103a8295b8c5dc4a7f2b&vid=be04d9d3e0a2b118e52f8d9224f8e9e527f6efa7&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR