Jasplakinolide ≥97% (by HPLC)

Supplier: Enzo Life Sciences

350-275-C050 350-275-C100
89149-388EA 282.31 USD
89149-388 89149-390
Jasplakinolide ≥97% (by HPLC)
Jasplakinolide

F-actin probe


Cell permeable F-actin probe. Induces actin polymerization. Completely inhibits the binding of phalloidin to F-actin. Antifungal and anti-proliferative agent.


Isolated from Jaspis splendens.

Formula: C₃₆H₄₅BrN₄O₆
MW: 709.7 g/mol
CAS Number: 102396-24-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=409f80d23f99065ee0b4d0f9c7ec2696bc0de137&vid=369ef465f05a3491c3f0d930a8c84e68a426be99&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR