Kahweol ≥97%

Supplier: Enzo Life Sciences

350-223-M010
89149-282EA 173.31 USD
89149-282
Kahweol ≥97%
Kahweol

GST activator.


  • Formulation: White to off-white solid.


Induces glutathione S-transferase. Has anti-cancer, chemoprotective, anti-inflammatory and anti-atherosclerotic properties.


Soluble in ethyl acetate, acetone or DMSO; insoluble in water.

Formula: C₂₀H₂₆O₃
MW: 314.2 g/mol
Storage Temperature: Freezer
CAS Number: 6894-43-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=789ff2c8387fa380612fb96e0ca0abcad3914b8a&vid=1030b64b20f96d9a860f8954cbef519c6f7405ab&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥97% (UPLC)
Source Natural product isolated from the unsaponifiable fraction of petroleum ether extract of coffee beans


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR