Dithiothreitol (DTT, Cleland's reagent) ≥98%

Supplier: Enzo Life Sciences

Synonyms: cis-2,3-Dihydroxy-1,4-butanedithiol, (R*,R*)-1,4-Dimercapto-2,3-butandiol, DTT, DL-1,4-Dithiothreitol

280-001-G005 280-001-G010 280-001-G025 280-001-G050
89148-136EA 124.26 USD
89148-136 89148-138 89148-140 89148-142
Dithiothreitol (DTT, Cleland's reagent) ≥98%
Dithiothreitol (DTT, Cleland's reagent)

Protective agent for SH groups


  • Formulation: White to off-white solid.


Cell permeable protective agent for SH groups, maintaining monothiols completely in the reduced state and of reducing disulfides quantitatively. ER-stress inducer.

Formula: C₄H₁₀O₂S₂
MW: 154.25 g/mol
MDL Number: MFCD00004877
CAS Number: 3483-12-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e1f28941be226ccf87f25412b0dae6ee44f5b2af&vid=c389b9f486112e105bacc5f7f98bbd8e853d7f90&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR