Anti-HSP90 Mouse Monoclonal Antibody (DyLight® 488)
Supplier: Enzo Life Sciences
SPA-830-488-F SPA-830-488-D
89106-716EA 673.62 USD
89106-716 89106-714
Anti-HSP90 Mouse Monoclonal Antibody (DyLight® 488)
Antibodies
The Hsp90 family of heat shock proteins represents one of the most abundantly expressed and highly conserved families of cellular chaperones whose expression can be upregulated under conditions of cellular stress, and includes cytoplasmic (Hsp90-alpha/beta), ER (grp94), and mitochondrial (TRAP1) localized members. Structurally, Hsp90 is characterized by an N-terminal ATP-binding domain, a medial substrate-binding domain, and a C-terminal dimerization motif. Hsp90 dimers function in cooperation with cochaperones (e.g. Hsp40, Hsp70, Hop, p23) to stabilize a multitude of client proteinMore Product Information