ActiveHOOK™ HyperCarrier™ Bovine Serum Albumin (BSA), Cationized (active), G-Biosciences
Supplier: G-Biosciences
786-097
82022-456EA 663.53 USD
82022-456
ActiveHOOK™ HyperCarrier™ Bovine Serum Albumin (BSA), Cationized (active), G-Biosciences
Proteins and Peptides
ActiveHOOK™ HyperCarrier™ is a pre‐activated form of HyperCarrier™ that has been activated by sulfoSMCC and is ready for direct coupling with peptides through the cysteine residues. HyperCarrier™ is a cationized form of bovine serum albumin. Cationized proteins are taken into antigen presenting cells (APC) quicker and more efficiently than normal proteins.  The positive charges on the cationized protein give it greater affinity for the negatively charged cell surface of the APCs. The cationized protein is internalized via activated receptor mediated endocytosis as opposed to pinocytosis (Border et al. (1982) J.Clin. Invest. 69:451). ActiveHOOK™ HyperCarrier™ is supplied lyophilized in 10mg/vial size.