Human Recombinant Caspase-9 (from E. coli)
Supplier: MilliporeSigma
218807-100U
80108-946EA 637.34 USD
80108-946
Human Recombinant Caspase-9 (from E. coli)
Proteins and Peptides
Member of the interleukin-1[beta] converting enzyme (ICE) family of cysteine proteases that exists in cells as an inactive proenzyme and is activated during the initiation of apoptosis
Functions to trigger the activation of downstream effector caspases and further leads to disassembly of cell structures. Specific activity: >=400units/mg protein.