TRIO.BAS™ AIRBIO ONE RAPID-VIRUS Sampling System
Supplier: Hardy Diagnostics
BAS2448K
76636-296EA 10575.33 USD
76636-296
TRIO.BAS™ AIRBIO ONE RAPID-VIRUS Sampling System
Samplers Air Samplers
The AIRBIO ONE RAPID-VIRUS System is an air sampler for airborne viable particle sampling. It is designed for total pathogen surveillance of bacteria, fungi, yeast, and viruses.
  • Collectionon unit is completely sterilizable
  • Tri-clamp stainless steel tube system facilitates cleaning and sterilization
Liquid sampling permits rapid methods (e.g. PCR), while traditional air sampling using culture media agar plates requires incubation of media plates to obtain sample results.
The dual-purpose AIRBIO ONE RAPID-VIRUS instrument may be used to collectMore Product Information