Medical Chemical Corporation Carbol Fuchsin, Ziehl-Neelsen, for AFB Stain for Mycobacteria, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
484A8
76591-146EA 31.22 USD
76591-146
Medical Chemical Corporation Carbol Fuchsin, Ziehl-Neelsen, for AFB Stain for Mycobacteria, Hardy Diagnostics
Stains Microbiology Stains
An aqueous solution of basic fuchsin, phenol and reagent alcohol. ‘Hot method’, requires heating of the slide.
Delivery information: Ship ground.