Orum International AIRBIO ONE Kit with Charging Cable, Hardy Diagnostics
Supplier: Hardy Diagnostics
BAS445K BAS446K BAS447K BAS448K
76509-302EA 9147.9 USD
76509-302 76509-304 76509-306 76509-308
Orum International AIRBIO ONE Kit with Charging Cable, Hardy Diagnostics
Samplers Air Samplers
The AIRBIO ONE instrument is both a stationary and portable air sampler with one aspirating head.
It is stable on a work surface without any support and ideally positioned for monitoring laminar airflow systems. Battery charges via wall charging cable. A second cable is included for optional transfer of sampling data to a PC with dedicated software (76446-766, separate purchase).
Ordering information: The kit includes one air sampler with power charger, one stainless steel aspirating head with cover, one data transfer cable, a calibration certificate, IQ, OQ, PQ fillable guidelineMore Product Information