HardyDisks™ AST Eravacycline, 20 µg (ERV20), Xerava, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
Z9401 Z9405
76501-650EA 17.14 USD
76501-650 76501-652
HardyDisks™ AST Eravacycline, 20 µg (ERV20), Xerava, Hardy Diagnostics
Microbiology Test Systems Microorganism Tests
HardyDisk™ AST’s are impregnated paper disks used for Antimicrobial Susceptibility Testing (AST); also known as disk diffusion or Kirby-Bauer testing.
Each cartridge contains 50 disks that are spring loaded for smooth and reliable dispensing.
Order Now
SPECIFICATIONS
Environmentally Preferable