Dermatophyte Milk Agar
Supplier: HARDY DIAGNOSTICS
L75
76359-394PK 76.64 USD
76359-394
Dermatophyte Milk Agar
Media for Microbiology Prepared Media for Microbiology
For the cultivation and differentiation of certain dermatophytes, especially Trichophyton rubrum, T. mentagrophytes, and Microsporum persicolor.