Mouse CCL28 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemical
KOA0399
76237-256EA 760.6 USD
76237-256
Mouse CCL28 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse CCL28 AccuSignal ELISA Kit
  • Natural and recombinant mouse CCL28.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: I23-R130
CCL28, also known as mucosae-associated epithelial chemokine(MEC), CCK1 and SCYA28, is a chemokine. It is mapped to 5p12. The tumor hypoxia promotes the recruitment of regulatory T(Treg) cells through induction of expression of the chemokine CCL28, which in turn promotes tumor tolerance and angiogenesis. CCL28 regulates the chemotaxis of cells that express the chemokine receptors CCR3 and CCR10. CCL28 has also been implicated in the migration of IgA-expressing cells to the mammary gland, salivary gland, intestine and other mucosal tissues. It has also been shown as a potential antimicrobial agent effective against certain pathogens, such as Gram negative and Gram positive bacteria and the fungus Candida albicans.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml mouse CCL28 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each mouse CCL28 standard solution and each sample be measured in duplicate.