Human CCL27 - CTACK ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemical
KOA0397
76237-252EA 760.6 USD
76237-252
Human CCL27 - CTACK ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human CCL27 - CTACK AccuSignal ELISA Kit
  • Natural and recombinant human CCL27.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: F25-G112
Chemokine(C-C motif) ligand 27(CCL27) is a small cytokine belonging to the CC chemokine family, it is also known under the names IL-11 R-alpha-locus chemokine(ILC), Skinkine, ESkine and Cutaneous T-cell-attracting chemokine(CTACK). This gene is mapped to 9p13.3. CCL27 is associated with homing of memory T lymphocytes to the skin, and plays a role in T cell-mediatedinflammation of the skin. CCL27 is expressed in numerous tissues, including gonads, thymus, placenta and skin. It elicits its chemotacticeffects by binding to the chemokine receptor CCR10.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml, 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml human CCL27 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human CCL27 standard solution and each sample be measured in duplicate.