Human MIP-1 alpha ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemical
KOA0257
76237-158EA 760.6 USD
76237-158
Human MIP-1 alpha ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human MIP-1 alpha AccuSignal ELISA Kit
  • Natural and recombinant human MIP-1 alpha.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence:  A27-A92
Macrophage inflammatory protein-1 alpha(MIP-1 alpha), also called CCL3, LD78. The cDNA for MIP-1 alpha predicts a mature peptide of 69 amino acids with a molecular mass of 7,889 daltons. LD78 is a member of a newly identified superfamily of small inducible proteins involved in inflammatory responses, wound healing and tumorigenesis. MIP-1 alpha is a chemokine that has pro-inflammatory and stem cell inhibitory activities in vitro. It constitutes an important second signal for mast cell degranulation in the conjunctiva in vivo and consequently for acute-phase disease. The standard product used in this kit is recombinant human MIP-1alpha, consisting of 66 amino acids with the molecular mass of 7.5kDa.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 15.6pg/ml human MIP-1 alpha standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA, citrate) to each empty well. It is recommended that each human MIP-1 alpha standard solution and each sample be measured in duplicate.