Tris HCl pH 9.0, 1.0 M
Supplier: Rockland Immunochemical
MB-028-1000
76236-404EA 92.99 USD
76236-404
Tris HCl pH 9.0, 1.0 M
Buffers Biological Buffers
Tris solution is an abbreviation of the organic compound known as tris(hydroxymethyl)aminomethane. Tris is extensively used in biochemistry and molecular biology.
  • This product is a concentrated stock solution and should be diluted appropriately with distilled, deionized water or equivalent to its final working concentration.
  • This buffer consists of 1.0 M Tris HCl at pH 9.0.
In biochemistry, Tris is widely used as a component of buffer solutions, such as in TAE and TBE buffer, especially for solutions of nucleic acids.
Application Note: Tris solution is suitable for molecular immunology, molecular biology and nucleic acid methodologies, when applicable. Visit our newly expanded web site at www.rockland-inc.com for methods using this and other buffers. This product is a concentrated stock solution and should be diluted appropriately with distilled, deionized water or equivalent to its final working concentration. This buffer consists of 1.0 M Tris HCl at pH 9.0.  Meticulously prepared using ultra pure reagents dissolved in highly polished pharmaceutical grade (WFI) water.