CYGREEN® Nucleic Acid Dye, Enzo
Supplier: Enzo Life Sciences
GEN105-0100
76003-286EA 163.5 USD
76003-286
CYGREEN® Nucleic Acid Dye, Enzo
Stains Electrophoresis Stains
Nucleic acid dye for gel staining and higher sensitivity qPCR.
  • Higher sensitivity allows detection of low target concentration
  • Greater PCR efficiency for accurate and reproducible results
  • Soluble in water for higher stability
  • Compatible with various enzyme systems
  • Suitable for commonly used rotor-type and plate-type qPCR platforms
  • Validated for qPCR applications and gel staining
  • Lower cost per assay compared to commonly used dyes
CYGREEN® Nucleic acid dye is a DNA intercalating agent that is used to stain DNA. The DNA-dye complex emits a fluorescence spectra that makes it suitable for qPCR and gel staining applications.CYGREEN® Nucleic acid dye has excitation/emission spectra similar to commonly used qPCR dyes and can be substituted in to increase sensitivity and reduce cost.