Anti-HMW Host Cell Proteins Rabbit Polyclonal Antibody
Supplier: Rockland Immunochemical
100-401-J06
75934-868EA 551.64 USD
75934-868
Anti-HMW Host Cell Proteins Rabbit Polyclonal Antibody
Antibodies
Anti-LMW Host Cell Proteins Antibody recognizes low molecular weight protein fractions from E.coli. This antibody has been anlyzed by 2D electrophoresis western blot to assure highest possible coverage of E.coli HCP. Host Cell Proteins are process-related impurities derived from a host cell expression system that may be present in trace amounts in a final drug substance. HCP Antibodies are used to detect impurities in biologically-based therapeutics. Detecting impurities often unseen by 1-D electrophoresis, a 2D electrophoresis western blot analysis delivers more complete insight into theMore Product Information