Swine Buffy Coat Preparation, Rockland Immunochemicals
Supplier: Rockland Immunochemical
R617-0100
75932-392EA 1144.03 USD
75932-392
Swine Buffy Coat Preparation, Rockland Immunochemicals
Blood
Swine Buffy Coat Preparation
  • aseptically collected and prepared to order
  • enriched for leukocytes (WBC)
Blood is a complex mixture of cells, proteins, metabolites and many other substances.  About 56% of human blood volume is comprised of cells, more than 99% of which are erythrocytes.  Human erythrocytes and thrombocytes (platelets, 0.5% blood components) do not contain nuclei.  Leukocytes (0.3% of blood components), also referred to as peripheral blood mononuclear leukocytes or PBMC, do contain nuclei.  Blood samples may varyMore Product Information