Regan-Lowe Semi Solid Agar Deep, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
Q32
75814-678PK 152.5 USD
75814-678
Regan-Lowe Semi Solid Agar Deep, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Regan-Lowe Semi Solid Agar Deep is used for the selective isolation of Bordetella pertussis.
  • Optically clear, shatter resistant tube
  • Selective Bordetella pertussis isolation
Charcoal Blood Agar with 10% Horse Blood and Cephalexin, for the selective isolation of Bordetella pertussis.