Water, deionised, sterile

Supplier: HARDY DIAGNOSTICS

Synonyms: Dihydrogen oxide

U284
75814-006PK 438.22 USD
75814-006
Water, deionised, sterile
Water

Sterile Deionized Water is useful for the preparation of dilutions and suspensions.


  • Created using the ion exchange process
  • Useful for general laboratory use
  • Lacks ions, such as those from sodium, calcium, iron and copper


Sterile deionized water was created using the ion exchange process and is a form of water which lacks ions, such as those from sodium, calcium, iron and copper. It is useful for general laboratory use and for the preparation of dilutions and suspensions.

Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100
Melting Pt: 0
MDL Number: MFCD00011332
CAS Number: 7732-18-5
Merck Index: 13,10098

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR