Anti-DNM1L Rabbit Polyclonal Antibody
Supplier: Rockland Immunochemical
600-401-AW7
10801-164EA 635.22 USD
10801-164
Anti-DNM1L Rabbit Polyclonal Antibody
Antibodies
The Dynamin-1-like protein (DNM1L) is a member of the dynamin superfamily of GTPases (1). DNM1L mediates mitochondrial and peroxisomal division, and is involved in developmentally regulated apoptosis and programmed necrosis (2). Dysfunction of this gene is implicated in several neurological disorders, including Alzheimer's disease. Mutations in this gene are associated with the autosomal dominant disorder, encephalopathy, lethal, due to defective mitochondrial and peroxisomal fission (EMPF) (3).
Recommended Dilutions: ELISA: 1:10,000-1:20,000; Immunofluorescence Microscopy: 20 ?g/mL;More Product Information