Anti-sTNF Receptor Type Rabbit Polyclonal Antibody (Biotin)

Supplier: Peprotech
500-P143BT-25UG
10776-384EA 339.8 USD
10776-384 10776-388
Anti-sTNF Receptor Type Rabbit Polyclonal Antibody (Biotin)
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human sTNF Receptor Type I. Anti­Human sTNF Receptor Type I­specific antibody was purified by affinity chromatography and then biotinylated.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Secondary
Antigen: sTNF Receptor Type
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR