Resorufinisobutyrate ≥97%

Supplier: ADIPOGEN CORP MS

Synonyms: 7-(Isobutyloxycarbonyloxy)-3H-phenoxazin-3-one

CDX-I0005-M001 CDX-I0005-M005 CDX-I0005-M025
102990-148EA 137.08 USD
102990-148 102990-150 102990-152
Resorufinisobutyrate ≥97%
Resorufinisobutyrate

Cell permeable resorufin derivative used as a fluorogenic indicator for cell viability. Resorufin was selected as the fluorophore because it fluoresces at a longer wavelength and is amenable to modification at the hydroxyl group which is essential for its fluorescence. Incubation with esterase at pH 8.0 results in a 80-90 nm shift of emission max. (Ex/Em: 500/~593nm in 0.1 M Tris pH 8.0 (after cleavage by esterase)). This cell viability indicator is 2 times more sensitive than calcein-AM.

Formula: C₁₇H₁₅NO₅
Storage Temperature: Freezer
CAS Number: 251292-24-7

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 97% (HPLC)
Appearance/Color Solid.
Solubility Soluble in DMSO.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR