AMCA-X ≥95%

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: 6-((7-Amino-4-methylcoumarin-3-acetyl)amino) hexanoic acid

CDX-A0073-M050 CDX-A0073-M250
102989-180EA 376.62 USD
102989-180 102989-182
AMCA-X ≥95%
AMCA-X

Amine-reactive blue fluorescent probe.

Formula: C₁₈H₂₂N₂O₅
Storage Temperature: Freezer
CAS Number: 205124-69-2

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 95% (HPLC)
Appearance/Color Solid.
Solubility Soluble in DMSO or acetonitrile.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR