Starch, red coloured

Supplier: ADIPOGEN CORP MS

CDX-S0035-G001 CDX-S0035-G005
102981-246EA 175.85 USD
102981-246 102981-248
Starch, red coloured
Starch

Red-dyed soluble starch. An excellent soluble substrate for alpha-amylase assay activity. Used for the measurement of enzyme activity, for research, forensic research, biochemical enzyme assays and in vitro diagnostic analysis.


  • Synthetic
  • Soluble in 0.5M potassium chloride solution

Formula: (C₆H₁₀O₅)n
MW: 162.14 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00082026
CAS Number: 9005-25-8

Order Now

Specification Test Results

Source/Host Synthetic
Appearance/Color Red or pink powder.
Solubility Soluble in 0.5M potassium chloride solution.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR