Human Recombinant Tumor Necrosis Factor Receptor Superfamily member 5
Supplier: Bon Opus Biosciences
C319-10UG C319-1MG C319-500UG C319-50UG
102861-326EA 57.08 USD
102861-326 102861-328 102861-330 102861-332
Human Recombinant Tumor Necrosis Factor Receptor Superfamily member 5
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human TNFRSF5 is produced by our Mammalian expression system and the target gene encoding Glu21-Arg193 is expressed with a 6His tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.