Human Recombinant Follistatin-Related Protein 1

Supplier: Bon Opus Biosciences

CC94-50UG CC94-10UG CC94-1MG CC94-500UG
102497-732EA 283.23 USD
102497-732 102508-946 102508-948 102508-950
Human Recombinant Follistatin-Related Protein 1
Proteins and Peptides

Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products


  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels


Recombinant Human Follistatin-like Protein 1 is produced by our Mammalian expression system and the target gene encoding Glu21-Ile308 is expressed with a 6His tag at the C-terminus.


Accessories information: Accessories not included.


Delivery information: Shipped Via FedEx


Packaging: Shipped at ambient temperature via FedEx envelope.


Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.

The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.

Order Now

SPECIFICATIONS

Protein/Peptide Name Follistatin-Related Protein 1
Environmentally Preferable
Protein/Peptide Type Recombinant
Species Human
Source

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR