C-Chip Disposable Hemacytometer, Bulldog Bio
Supplier: Bulldog Bio
DHC-N005 DHC-N01 DHC-N002
102407-944EA 75 USD
102407-944 102407-946 102408-108
C-Chip Disposable Hemacytometer, Bulldog Bio
Slides Counting Chambers
The C-Chip offers the ability to eliminate tedious washing and reduce exposure to infectious samples and contamination
In a conventional glass hemocytometer, improper fitting of the chamber and coverslip changes the volume of the sample introduced into the chamber. The C-Chip’s all-in-one “chamber with an integrated cover” solves the problem. For general, mammal, erythrocyte, and leukocyte cell counting. Individually sealed.
Certifications: Lot tested and certified with certificate in box.