PUREXPRESS® RF123 Kit, New England Biolabs
Supplier: New England Biolabs (NEB)
E6850S
101641-564EA 441 USD
101641-564
PUREXPRESS® RF123 Kit, New England Biolabs
Protein Expression Protein Expression Systems
Release factors are involved in termination of protein translation by recognizing the stop codons in an mRNA sequence
In a ribosome display experiment using PURExpress, lack of release factors could stabilize the ternary complex of mRNA-ribosome-nascent protein. As a result, the cDNA recovery could be higher. In this kit, the three release factors are supplied separately, allowing users to perform a protein synthesis reaction/ribosome display experiment with/without release factors of their choice.
Order Now
SPECIFICATIONS
Environmentally Preferable