Anti-PIP2 Mouse Monoclonal Antibody [clone: NOT AVAILABLE]
Supplier: Enzo Life Sciences
915-052-020
101106-530EA 334.63 USD
101106-530
Anti-PIP2 Mouse Monoclonal Antibody [clone: NOT AVAILABLE]
Antibodies
Metabolism of inositol phospholipids by intracellular signaling mediators is fundamental to signal transduction in eukaryotic cells. PI-4,5-P2 (PIP2) can be synthesized by phosphorylation of PI-4-P by type I phosphatidylinositol phosphate kinase (PIP5K I), or phosphorylation of PI-5-P by type II PIPK (PIP4K II). PI-4,5-P2 regulation of cellular calcium levels involves its hydrolysis by Phospholipase C (PLC) to produce inositol 1,4,5-triphosphate (IP3) and diacylglycerol (DAG), which serve as second messengers in the import of calcium via IP3-sensitive ion channels and in the activation ofMore Product Information