IHC Diluent (Primary Ab), Enzo Life Sciences
Supplier: Enzo Life Sciences
950-244-0250
10051-956EA 156.96 USD
10051-956
IHC Diluent (Primary Ab), Enzo Life Sciences
Stains Microbiology Stains
IHC diluent (primary Ab) can be used as a diluent to prepare primary antibodies and negative controls
Liquid. Primary antibody diluent.
It can be used with any primary antibody irrespective of its source of origin. Primary antibody diluent is specifically designed to stabilize primary antibodies and reduce nonspecific background staining observed during immunostaining.