VICTOR® Nivo™ Multimode Plate Readers, PerkinElmer
The VICTOR Nivo system is a high-performance filter-based multimode microplate reader equipped with all major detection technologies:
- Absorbance
- Luminescence
- Fluorescence intensity
- Time-resolved fluorescence
- Fluorescence polarization
- Alpha
It’s a compact, light-weight multimode plate reader designed for life science research laboratories performing routine low-throughput assays, or assay development work, with diverse application requirements. When combined with the stacker, throughput for the VICTOR Nivo is increased and productivity is enhanced by enabling users to set up walk away automation so they can focus on their research and not on managing plates.
The VICTOR NIVO system’s software has a modern, workflow-oriented user interface which is easy to learn and use and includes pre-written application protocols to get users productive quickly. In addition, MyAssays Desktop Standard software is provided for data analysis. With our Enhanced Security software option, it contains technological tools to help guide you through the complexity of compliance for 21 CFR Part 11.
The browser-based control software is independent of operating system, so it can be controlled via a variety of touchscreen devices, for example Windows or Mac OS PCs or tablets.
The VICTOR Nivo microplate reader can be operated via WiFi or a network connection, enabling experiments to be set-up, run and monitored remotely, such as from outside a biosafety cabinet or from a neighbouring lab.
Victor Nivo 3T Multimode Plate Reader
This system provides: luminescence, absorbance, and fluorescence. The VICTOR® Nivo™ system packs all popular detection modes into the industry’s smallest multimode reader footprint. The perfect microplate reader for everyday biochemical and cell-based as
Victor Nivo 5T Multimode Plate Reader
This system provides: luminescence, absorbance, fluorescence, time resolved fluorescence, and fluorescence polarization. The VICTOR® Nivo™ system packs all popular detection modes into the industry’s smallest multimode reader footprint. The perfect micro
Victor Nivo 6T Multimode Plate Reader
This system provides: luminescence, absorbance, fluorescence, time resolved fluorescence, fluorescence polarization, and Alpha. Now with Alpha Technology, the VICTOR® Nivo™ system packs all popular detection modes into the industry’s smallest multimode re
Accessories for Victor Nivo Multimode Plate Readers, PerkinElmer
Designed exclusively for PerkinElmer VICTOR® Nivo™ Multimode Plate Readers