Histology Laboratory Consumables
Proper handling of sample materials by utilizing high-quality histology consumables will save time and reduce waste. These consumable products support the production of clean and accurate results, helping advance biological research and development. With so many histology applications in medicine, science, and education, it’s important to understand which histology techniques are best suited for your laboratory. Select from specialized histology consumables and equipment for each step of the preparation workflow, from specimen collection to finished slides ready for examination.
Featured products
Embedding Cassettes
Choose from various shapes, sizes, and features that accommodate any specimen. Options for different cassette colors, large labeling areas, and printer compatibility make for effortless identification.
Embedding Stations
For processing a larger number of samples, Avantor® offers embedding stations that streamline sample preparation for research applications. Regulate temperature and the placement of specimens for precise ribbon sectioning for accurate and reproducible res
Microtomes, Cryostats, and Blades
Precise, thin cutting is essential for microscopic examinations. Select from a variety of microtomes and blades ideal for any specimen or hardness. Cryostats keep your materials at a controlled temperature so you can focus on research and development.
Paraffin
Used to impregnate tissue prior to sectioning thin samples, paraffin is a staple in histology sample preparation and handling. Keep your lab well stocked with our trusted products.
Tissue Processors
Automated tissue processors operate in a fraction of the time manual procedures require. Upgrade your lab with either benchtop or floor model tissue processors.
Water Baths
An integral part of histology procedures, water baths are designed to assist with handling paraffin wax samples. Handle your samples with easy-to-set level controls, and sensors always guarantee temperature uniformity.
What are the applications of histology?
Histology is a versatile scientific study examining the microanatomy of cells and tissues. The sectioning and staining of the tissues can provide vivid imagery of samples, making it possible for researchers to understand the biological processes of a tissue or cell. Applications of histology include:
- Aid in disease diagnosis
- Medical innovations and discovery
- Education and research (ex: Archaeology)
- Forensic analysis In all these fields, having quick and dependable access to consumables is needed to prepare samples and run a histology report effectively. In many cases, laboratory protocols will call for a large quantity of samples to be tested making it critical to optimize your methods to save time and resources.
2 Ways to optimize histology sample preparation with lab consumables
Proper handling of your samples and laboratory supplies can ensure readable and consistent results. Dependable protocols can take considerable time and effort to establish, making it easy to waste consumable materials and sample tissue in the process. There are ways you can prepare your workspace and team to minimize errors and waste when operating a histology lab.
Handle histology samples with care
When it comes to successful histology testing, the correct handling of your sample reduces waste and supports viable results. Cytology samples can be significantly easier to work with if you try the following:
- Use microscope slides with frosted ends
- Rapidly drying your slides after preparation results in superior cell preservation
- Handling tissue samples in separate containers will prevent contamination or wasted materials.
- Fix tissues by using the correct ratio of formalin and sample tissue (10:1 by volume)
Properly prepare and embed your sample tissue
Before sectioning your sample in the microtome, the tissue sample must be embedded. This is done by immersing the sample in molten paraffin wax or frozen in a solution. This process is key for preserving biological materials in the sample and will aid in preserving both tissue morphology and providing a clean cut during the sectioning phase. Once a sample is set, use caution when opening lids, as tissue often sticks to them. In cases where tools need to be used, it is good practice to keep your equipment sanitized to avoid any contamination at this phase. Beyond the handling of sample material, selecting the correct histology consumables will provide a foundation for successful bioprocessing. This starts with choosing the right containers and specifically, cassettes that are compatible with your sample materials.
The difference between biopsy cassette and tissue cassette
Cassettes are essential for the processing of tissue blocks in a histology lab. Both biopsy and tissue cassettes provide a similar mechanism for histology testing. They are made up of materials resistant to histological solvents and are both designed with slots to offer maximum flow through. This enables good fluid exchange and proper drainage from the cassette. The key difference between these two histology lab consumables is the design of the slots in the cassettes. Biopsy cassettes feature a small grid of circular slots, whereas tissue cassettes have longer slot openings within the tray. This distinction in design is due to the nature of how each type of tissue is processed. For a biopsy test, the tissue samples are typically small and benefit from being passed through a 1mm square matrix. Depending on the thickness and state of the sample sections being tested, having proper flow will support consistent results and avoid any degradation of your sample in the process. Once you have identified the best cassettes for your process, you will need to make sure that both your cassettes and sample materials are processed through the correct microtome. This selection process comes down to two fundamental details: what materials will you be cutting into sections, and what is the best blade to meet your needs?
7 Common types of microtome blades
There are several options when deciding the best microtome blades for your process. Disposable microtome blades come in different sizes and materials – each designed to slice through various types of material. Your selection relies heavily on the type of microtome equipment you use. To understand more about the specifications of each type of microtome blade, here is a comprehensive guide to the seven most common types of blades used.
1. Rotary microtome blade
Rotary microtomes require a slightly heavier knife, usually measuring between 0.5 to 60 µm. These systems are great for cutting semi-thin to thin sections for light microscopy. They are designed with a hand-wheel operated blade, with the option to modify the angle of the knife to cut larger blocks of tissue as needed.
2. Sledge microtome blade
Sledge microtome blades are significantly larger, measuring around 24cm in length with a wedge shape to minimize vibration. This makes these blades ideal for cutting larger blocks of tissue compared to other machines. A sledge microtome works by setting the tissue block on a steel carriage, which then slides back and forth over a fixed blade. Sledge microtome blades are perfect in applications like segmenting of an entire brain or other large organ tissues.
3. Vibrating microtome blade
A common tool of histochemistry, vibrating microtomes were developed to section fresh plant or animal materials for studying. It operates with a high-speed vibration, aiding in the cutting of soft materials immersed in fluid. This process requires disposable, double-edged razor blades; however, some specialized knives are available.
4. Ultramicrotome blade
Ultramicrotome blades are known for their ability to cut incredibly thin sections. This is perfect for both light and electron microscopy. These blades are made from materials like glass, diamond, or sapphire in order to maintain a very sharp edge for uniform, ultra-thin cuttings.
5. Laser microtome blade
In contrast with the other microtome blades, this process does not utilize a physical blade for cutting sections of tissue. Rather, laser microtomes are built with a bladeless femtosecond laser, to produce samples with great precision. Microtome lasers can produce sections with a thickness of around 5 to 100µm. This method works well on biological materials and a range of other materials.
6. Saw microtome blade
For harder materials, such as bone, ceramic, or resin-embedded samples, saw microtomes are the best option for processing your sample. Materials are carefully cut using a rotating diamond-coated saw. There are some limitations for the sizing of sections, as you will not be able to produce sections smaller than 20µm with this kind of blade.
7. Cryostat blade
Cryostat blades were designed to cut thin sections of frozen tissue. A cryostat is built as a deep-freeze cabinet able to house a rustproof microtome. These systems are compatible with a multitude of blades depending on both the microtome model and the materials that will be cut and sectioned.
Conclusion
Histology laboratory consumables make handling your biological samples simple. Refine your laboratory techniques for success - saving time and valuable resources by utilizing all of the histology consumables we have to offer. Select the ideal vessels, hardware, and disposable products to meet your unique needs. Whether you are studying biological results from an archeological laboratory or discovering new medical implications of a drug – having dependable equipment is necessary. Learn more about all the products and resources for histology through Avantor®.