Specialty Silver Products
Some of the key features of our silver products include: silver compounds are manufactured up to a purity of 99.999%, a batch specific certificate of analysis is produced for each product, each silver product batch has full metallic impurities typically measured by ICP-MS to ppm and measured silver content.