SW Technologies
SW® invests in manufacturing, glove performance, and hand health technologies to provide premium products that protect and enhance worker performance.
MANUFACTURING – SW has focused on developing new manufacturing technologies to drive category innovation and improve worker performance.
HAND HEALTH – SW proprietary technologies that improve hand health and counter the effects associated with wearing hand protection.
GLOVE PERFORMANCE – SW has developed a number of new technologies that have transformed the single-use glove category – from simply protecting hands to enhancing comfort, enabling better performance, to protecting the environment.
pH Natural®
Actively maintains skin pH at natural levels to reduce skin issues. Proprietary coating helps keep pH levels at 5.5, reducing microbial growth.
Dry Core®
Naturally pulls sweat away from the skin for healthier hands. Moisture-wicking system makes for a dry, comfortable environment.
EcoTek®
Available in many popular PowerForm®, TrueForm®, and MegaMan® single-use styles, EcoTek is SW’s patented, environmentally sustainable nitrile glove formula. EcoTek is manufactured sustainably and rigorously tested per the ASTM D5526 standard*. EcoTek is useful for all glove users and industries as it reduces emissions when incinerated.
TracTek®
Proprietary matrixed pattern that provides dramatic control. Designed for maximum surface-to-surface contact, providing enhanced grip in all conditions.