SHEL LAB B.O.D. Thermoelectric Cooled Incubators
SHEL LAB’s B.O.D. Thermoelectric Cooled Incubator use innovative thermoelectric cooling technology, the SRI6P and SRI20P eliminate the need for a refrigeration compressor. This means that these units use 78% less power than alternative models and reduce room air conditioning loads by 75%. They also include 75-pound capacity shelves, which eliminates sagging.
SRIP incubators meet APHA specifications for Biochemical Oxygen Demand (BOD) and include a mechanical convection system to ensure even air distribution, digital temperature set controller, over-temperature limit control, and a digital temperature display
SRI6P B.O.D Incubator - 185 L
SHEL LAB SRIP Incubators excel at biological oxygen demand (BOD) applications.
SRI20P B.O.D Incubator - 546 L
SHEL LAB SRIP Incubators excel at biological oxygen demand (BOD) applications.