Agar
Highest Quality, Widest Selection for Every Application
Prepared in our on-site labs by experienced biologists and lab technicians, we have the widest selection of prepared media in a variety of packaging options to suit your needs.
Agar Packaging Options*
Tubes
Set of twelve 16 x 125mm screw cap tubes. Slanted, unless noted.
Media Pour Pack
Package of 20 sterile Petri plates (90 x 15mm) and three media bottles. Each bottle pours seven plates.
Plates
Ten 90 x 15mm plates, sealed. Can be stored for up to two weeks.
Bottle
Six 125mm bottles, sufficient for 15-20 tubes or 5-7 plates per bottle.
*Available options vary by individual product. See product ordering page to select from the packaging options available for that particular item.