High Purity Water
Put the simplicity back into water with RICCA as the premier choice for all your water needs. We offer the broadest line of water types, packaging and testing configurations. RICCA's state of the art water and quality systems at multiple facilities means rapid manufacturing and delivery to anywhere in North America. With the tightest specifications in the industry, you can expect only the best.
- USP Purified, Sterile Filtered, WFI Quality
- LC/MS
- HPLC
- ACS Reagent Grade, ASTM Type I, II, III, and IV
- USP/EP/JP Purified
- Distilled
- Sterile, Molecular Biology Grade
Product selection
USP Sterile Purified Water, WFI Quality
Suitable for pharmaceutical, bio-processing, medical device manufacturing and research applications.
LC/MS Water
With Low UV Absorptivity, RICCA LC/MS water is suitable for critical applications across all wavelengths.
HPLC Water
For use in LC, HPLC and UPLC applications.
ACS/ASTM Water
Deionized water that meets ACS and ASTM Type I-IV Specifications.
USP/EP/JP Purified Water
Ideal for pharmaceutical applications, our USP, EP, and/or JP purified water has low microbial count to guarantee quality with use.
Sterile, Molecular Biology Grade Water
Free of RNase, DNase, Protease, DEPC, and other chemical additives.