Other PPE & Sterilization Wrap
The HALYARD* PPE portfolio offers a full line of protective apparel for both staff and patients. In many cases, our protective apparel meets or exceeds performance standards and guidelines established by organizations such as OSHA, AAMI, NIOSH, and ASHP for strength, barrier protection, and fluid resistance. HALYARD* Sterilization Wrap patented DAISY DESIGN* Pattern is engineered to optimize the surface area available for sterilant penetration and evacuation. 
*†The DAISY DESIGN is a Registered Trademark of Kimberly-Clark Worldwide, Inc. Used under license.
Sequential Sterilization Wrap
HALYARD* Sequential Sterilization Wrap is the fast, easy way to ensure instrument sterility is uncompromised.
BASIC PLUS* Lab Coats
HALYARD* BASIC PLUS* Lab Coats all feature a protective 3-layer fabric that offers exceptional resistance to blood strike-through at various levels of exposure.
FLUIDSHIELD* Surgical N95 Respirator Mask
FLUIDSHIELD* N95 filter respirators are NIOSH approved and are designed to provide an effective facial fit as required for proper respiratory protection.
HALYARD* CHEMO 360* Procedure Gown
Gear up for the fight now because there is no safe level of exposure to cytotoxic chemotherapy drugs for healthcare workers. Choose the HALYARD* CHEMO 360* Chemotherapy Procedure Gown to protect against 52 chemotherapy drugs.