Pipettes
Our pipette line includes many application-specific varieties: bacteriological, serological, and measuring pipettes to name a few. In addition, we offer different characteristics to suit specific purposes including glass, plastic, and electronic versions plus a variety of sizes.
Pasteur Pipettes
Choose from pipettes guaranteed to accurately transfer or measure liquid samples.
Volumetric Pipettes
DWK volumetric Pipettes are manufactured according to both ASTM E543, and E969, are NIST traceable and will provide years of accurate service.
Bacteriological Pipettes
Glass Pipettes that are designed specifically for testing milk and milk products for bacterial contamination
Serological Pipettes
Available in sterile and non-sterile, disposable serological pipettes are manufactured from 51-expansion borosilicate glass.
Automatic and Manual Pipettes
DWK's complete line of pipettes offers your lab a single source for superior quality pipettes