High Purity Reagents and Solvents for Inorganic Trace Analysis
The Honeywell TraceSELECT™ product range includes high purity acids, bases, solvents and salts that are designed for trace and ultra-trace analysis.
Sample preparation for inorganic trace analysis applications requires reagents of the highest purity. Specifically, they must have very low metal content to ensure appropriately low blanks. Our reagents are handled under clean-room conditions and supplied in high quality leach resistant bottles to ensure long-term purity. The certificates of analysis list up to 80 individual metal and anion traces.
Product selection
TraceSELECT Ultra Acids and Bases
For the most demanding ultra-trace analysis at parts per billion (ppb) and parts per trillion (ppt) levels.
TraceSELECT Acids, Bases and Salts
Economical reagents for trace analysis in the ppb level with typical blank levels less than 1 ppb.
TraceSELECT Solvents
High purity solvents for speciation analysis by LC-ICP-MS.
TraceSELECT Reagents
Reagents for Atomic Absorption Spectroscopy (AAS) and Voltammetry (VA) such as matrix modifiers, spectroscopic buffers, reducing agents and additives.