Tube Racks
We offer the largest portfolio of classic, adaptable and interlocking racks.
OneRack® Test Tube Racks
These full size Derlin® (fiberglass-reinforced polyoxymethylene) racks offer the highest chemical and heat resistance. All racks feature molded coordinates to facilitate tube orientation and selection. They are designed to maintain stable use in water baths and include a flat labeling area for quick and easy identification. Racks are autoclavable.
HDPE Coated Wire Racks
Chemical resistant racks are durable for transport or sample handling. 
4-Way Microtube Rack
Heavy-duty polypropylene racks feature a unique system of tabs and slots, which facilitate easy connection and sturdy fit. Each rack can hold four 50 mL conical tubes, twelve 15 mL conical tubes, thirty-two 1.5 mL microtubes, or thirty-two 0.5 mL microtubes. Racks are autoclavable. Assorted color rack orders include one each of blue, green, pink, yellow, and orange.
96-Well PCR Rack
Sturdy polypropylene rack accommodates individual 0.2 mL tubes or strips of eight or twelve tubes. Tube wells are easily identified with embossed numbers and letters. The removable hinged lid lies flat. Racks are autoclavable.